Learn More
Crystal Chem Inc Human Creatinine Assay Kit, 5-Kit Pack, 96-tests per kit
SDP
Catalog No. 501947671
Encompass_Preferred
The Creatinine Assay is used to quantify levels of creatinine in human serum and urine in less than 10 min. The 5-Kit Pack is convieniently provided as five individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight (8) µL of sample, ideal for research applications. Designed for human samples.Crystal Chem is a worldwide leading supplier of ELISA kits for human research makers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7671 Supplier Crystal Chem Inc Supplier No. 80185
The Creatinine Assay is used to quantify levels of creatinine in human serum and urine in less than 10 min. The 5-Kit Pack is convieniently provided as five individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight (8) µL of sample, ideal for research applications. Designed for human samples.Crystal Chem is a worldwide leading supplier of ELISA kits for human research makers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.