Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CREB3L4 (aa 265-370) Control Fragment Recombinant Protein

Catalog No. RP89174
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89174 Supplier Invitrogen™ Supplier No. RP89174
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58254. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TEY5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 148327
Name Human CREB3L4 (aa 265-370) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40
Common Name CREB3L4
Gene Symbol CREB3L4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELERHNISLVAQLRQLQTLIAQTSNKAAQTSTCVLILLFSLALIILPSFSPFQSRPEAGSEDYQPHGVTSRNILTHKDVTENLETQVVESRLREPPGAKDANGSTR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.