Learn More
BIOHIPPO Human CREBBP Microsample
SDP
Catalog No. 5000314510
Encompass
Human CREBBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CREBBP in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CREBBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CREBBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CREBBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14510 Supplier BIOHIPPO Supplier No. BHE15211630
Human CREBBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CREBBP in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CREBBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CREBBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CREBBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color