Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CRELD2 ELISA Kit

Catalog No. EH128RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH128RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH128RBX10 Supplier Invitrogen™ Supplier No. EH128RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human CRELD2 quantitates human CRELD2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRELD2.

CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UXH1
Assay Range 0.137-100 ng/mL
Assay Sensitivity 0.13 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 5730592L21Rik; C85758; CREL2; CRELD2; cysteine rich with EGF like domains 2; cysteine rich with EGF-like domains 2; cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2; RGD1310614; UNQ185/PRO2115730592L21Rik; C85758; CREL2; CRELD2; cysteine rich with EGF like domains 2; cysteine rich with EGF-like domains 2; cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2; RGD1310614; UNQ185/PRO211;
Gene ID (Entrez) 79174
Gene Symbol CRELD2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CRELD2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.