Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CRIM1 ELISA Kit

Catalog No. EH129RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH129RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH129RBX10 Supplier Invitrogen™ Supplier No. EH129RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human CRIM1 quantitates human CRIM1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRIM1.

CRIM1 (cysteine-rich motor neuron 1), a glycosylated type I transmembrane protein, plays a role in tissue development i.e. capillary formation and maintenance during angiogenesis. It contains an N-terminal IGF-binding protein-like motif and six von Willebrand-like cysteine-rich repeats (CRRs) in its extracellular domain. CRIM1 interacts with BMP4 and BMP7 via the CRRs and functions as an antagonist. CRIM1 is developmentally expressed in a number of tissues including the pancreas, kidney, placenta, brain and blood vessels. CRIM1 may participate in CNS and placental development by interacting with growth factors involved in motor neuron differentiation and survival.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZV1
Assay Range 0.041-10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330;
Gene ID (Entrez) 51232
Gene Symbol CRIM1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CRIM1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.