Learn More
Raybiotech Inc Human Cripto-1 ELISA Kit
Supplier:  Raybiotech Inc ELHCRIPTO11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cripto-1.
Catalog No. 50-194-9152