Learn More
Biomatik Corporation Human Crk Like Protein (CRKL) ELISA Kit
Supplier:  Biomatik Corporation EKU03499
Encompass_Preferred
Human Crk Like Protein (CRKL) ELISA Kit for detecting CRKL in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46109. Gene ID: 1399. Target Synonyms: V-Crk Sarcoma Virus CT10 Oncogene Homolog (avian)-Like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5226