Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CRMP3 (aa 499-535) Control Fragment Recombinant Protein

Catalog No. RP100153
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100153 Supplier Invitrogen™ Supplier No. RP100153
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83947. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Collapsin response mediator proteins (CRMPs), including CRMP-1 (DRP-1), CRMP-2 (DRP-2 or TOAD64), CRMP-3 (DRP-4), CRMP-4 (DRP-3) and CRMP-5 (DRP-5), mediate signal transduction after exposure of neural cells to the axon guidance molecule Semaphorin 3A (SEMA3A)/collapsin. CRMPs are present in the developing cerebral cortex and neocortical neurons and are responsive to SEMA3A. In the adult brain, the expression of CRMPs is dramatically downregulated. However, they remain expressed in structures that retain their capacity for differentiation and plasticity. CRMP-3 is expressed transiently in developing spinal cord and selectively in the postnatal cerebellum.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14531
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 10570
Name Human CRMP3 (aa 499-535) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI173505; Collapsin response mediator protein 3; CRMP3; CRMP-3; dihydropyrimidinase like 4; dihydropyrimidinase-like 4; dihydropyrimidinase-related protein 4; DPY4; Dpys4; DPYSL4; DRP-4; ULIP4; ULIP-4; ULIP4 protein; unc-33-like phosphoprotein 4; UNC33-like phosphoprotein 4
Common Name CRMP3
Gene Symbol DPYSL4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YDGPVHEVMVPAKPGSGAPARASCPGKISVPPVRNLH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.