Learn More
Biomatik Corporation Human Cross Linked C-Telopeptide Of Type III Collagen (CTXIII) ELISA Kit
Supplier:  Biomatik Corporation EKU03508
Encompass_Preferred
Human Cross Linked C-Telopeptide Of Type III Collagen (CTXIII) ELISA Kit for detecting CTXIII in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 46.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02461. Gene ID: 1281. Target Synonyms: CTX-III; CTX3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4740