Learn More
Abcam Human CRP ELISA Kit (C-Reactive protein), 10 x 96 Tests
Catalog No. 502656764
Encompass_Preferred
Human CRP ELISA Kit (C-Reactive protein) is a single-wash 90-min Simplestep used to quantify Human CRP (C-Reactive protein) with a sensitivity of 5.36 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6764 Supplier Abcam Supplier No. AB26005810X96TESTS
Add to Cart
Add to Cart
Request a Quote
Human CRP ELISA Kit (C-Reactive protein) is a single-wash 90-min Simplestep used to quantify Human CRP (C-Reactive protein) with a sensitivity of 5.36 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement