Learn More
Sino Biological Recombinant Anti-C-Reactive Protein Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 11250R106H20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human C-Reactive Protein/CRP (rh C-Reactive Protein/CRP; Catalog#11250-H08H; NP_000558.2; Met1-Pro224), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4750