Learn More
Sino Biological Recombinant Human C-Reactive Protein Protein, Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 11250HNAHB20
Encompass_Preferred
A DNA sequence encoding the human CRP (P02741-1) (Met1-Pro224) was expressed and purified. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4751