Learn More
Sino Biological Recombinant Human C-Reactive Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11250H49HB20
Encompass_Preferred
A DNA sequence encoding the Human CRP (P02741-1)(Met1-Pro224) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1107