Learn More
Invitrogen™ Human CRTAC1 ELISA Kit
Description
Human CRTAC1 quantitates human CRTAC1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRTAC1.
Specifications
Specifications
| Accession Number | Q9NQ79 |
| Assay Range | 1.23-300 ng/mL |
| Assay Sensitivity | 1.23 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | 2810454P21Rik; 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; ASPIC1; AW047536; cartilage acidic protein 1; Cep68; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; Crtac1; Crtac1B; FLJ10320; Lotus; Protein CRTAC1-B; W3072810454P21Rik; 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; ASPIC1; AW047536; cartilage acidic protein 1; Cep68; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; Crtac1; Crtac1B; FLJ10320; Lotus; Protein CRTAC1-B; W307; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.