Learn More
Raybiotech Inc Human CRTAM ELISA Kit
Supplier:  Raybiotech Inc ELHCRTAM1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ACE-2, Albumin, AMICA, ANG-4, BAFF, CA19-9, CD163 , Clusterin, CXCL14, Cystatin C, Decorin, Dkk-3, DLL1, Fetuin A, aFGF, FOLR1, Furin, GASP-1, GASP-2, GCSF R, HAI-2, IL-17B R, IL-27, LAG-3 , LDL R , Pepsinogen I, RANK, RBP4, SOST, Syndecan-1, TACI, TFPI, TSP-1, TRAIL R2 , TRANCE, Troponin I, uPA, VE-Cadherin, WISP-1.
Catalog No. 50-194-9158