Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio IVT BIOIVT LLC Human Mononuclear Cells, Cryopreserved, Individual Donor, High Viability

Catalog No. NC4000090
Encompass

Human Mononuclear Cells (PBMC) are provided cryopreserved from individual donors with high viability. This product is intended for in-vitro research and manufacturing purposes only, requiring handling at Bio-Safety Level 2.

  • HLA type provided for detailed donor characterization.
  • Tested negative for a panel of infectious agents including HIV ½ AB, HCV AB, HBSAG, HIV-1 RNA, HCV RNA, HBV DNA, WNV RNA, ANTI-T. CRUZI, and STS.
  • Recommended short-term storage at -70°C or colder for up to 2 weeks.
  • Recommended long-term storage in liquid nitrogen vapor phase (-140°C).
  • Sample type from individual male donor.
  • Donor race is white or Caucasian.
  • Concentration per vial is 10M.
  • Viable cell count of 20.0 × 106.
  • Cell viability is 91.0%.
  • Cryopreserved using CRYOSTOR CS10.

Catalog No. NC4000090 Supplier Bio IVT Supplier No. HUMANPBMC0002021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Mononuclear Cells (PBMC) are provided cryopreserved from individual donors with high viability. This product is intended for in-vitro research and manufacturing purposes only, requiring handling at Bio-Safety Level 2.

  • HLA type provided for detailed donor characterization.
  • Tested negative for a panel of infectious agents including HIV ½ AB, HCV AB, HBSAG, HIV-1 RNA, HCV RNA, HBV DNA, WNV RNA, ANTI-T. CRUZI, and STS.
  • Recommended short-term storage at -70°C or colder for up to 2 weeks.
  • Recommended long-term storage in liquid nitrogen vapor phase (-140°C).
  • Sample type from individual male donor.
  • Donor race is white or Caucasian.
  • Concentration per vial is 10M.
  • Viable cell count of 20.0 × 106.
  • Cell viability is 91.0%.
  • Cryopreserved using CRYOSTOR CS10.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.