Learn More
Bio IVT BIOIVT Human Mononuclear Cells (PBMC), 20x10^6 Cells/Vial, Cryopreserved, Gender Unspecified
Catalog No. NC4000092
Encompass_Preferred
This product consists of Human Mononuclear Cells (PBMC) cryopreserved in CryoStor CS10 (LMX ACD A) solution. Each vial contains 20 x 106 cells. These cells are manufactured using optimized methods to ensure high quality with superior viability, purity, and functionality.
  • Contains 20 x 106 cells per vial.
  • Cryopreserved in CryoStor CS10 (LMX ACD A).
  • Manufactured with optimized methods for superior viability, purity, and functionality.
  • Reduces sample preparation time.
  • Available cryopreserved for future scheduled work.
  • Prepared from various source matrices, including vacutainer collection, buffy coat, and leukopak.
  • Donors provide informed consent under IRB-approved protocols.
  • Samples are fully characterized with demographic and medical data.
  • Donor gender is unspecified.
Catalog No. NC4000092 Supplier Bio IVT Supplier No. HUMANPBMC0101311
This product consists of Human Mononuclear Cells (PBMC) cryopreserved in CryoStor CS10 (LMX ACD A) solution. Each vial contains 20 x 106 cells. These cells are manufactured using optimized methods to ensure high quality with superior viability, purity, and functionality.
  • Contains 20 x 106 cells per vial.
  • Cryopreserved in CryoStor CS10 (LMX ACD A).
  • Manufactured with optimized methods for superior viability, purity, and functionality.
  • Reduces sample preparation time.
  • Available cryopreserved for future scheduled work.
  • Prepared from various source matrices, including vacutainer collection, buffy coat, and leukopak.
  • Donors provide informed consent under IRB-approved protocols.
  • Samples are fully characterized with demographic and medical data.
  • Donor gender is unspecified.