Learn More
Raybiotech Inc Human Cryptic ELISA Kit
Supplier:  Raybiotech Inc ELHCRYPTIC1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cryptic.
Catalog No. 50-194-9159