Learn More
Sino Biological Recombinant Anti-M-CSF/CSF1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11792R033100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human M-CSF / CSF-1 (rh M-CSF / CSF-1; Catalog#11792-H08H; P09603-1; Met1-Asn190).
Catalog No. 50-241-0947