Learn More
Sino Biological Recombinant Anti-M-CSF/CSF1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11792R028100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human M-CSF/CSF-1 (rh M-CSF/CSF-1; Catalog#11792-HNAH; NP_757349.1; Met1-Asn190).
Catalog No. 50-241-1139