Learn More
Sino Biological Recombinant Human M-CSF/CSF1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11792H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CSF1 (P09603-1) (Met1-Arg255) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4550