Learn More
Sino Biological Recombinant Human M-CSF/CSF1 Protein (His Tag)
Supplier:  Sino Biological 11792H08H1
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Met 1-Asn 190) of human CSF1 (P09603-1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4553