Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10161MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CSF1R (rh CSF1R; Catalog#10161-H08H; NP_005202.2; Met1-Glu512). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0084