Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10161T58100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human M-CSFR/CSF1R (rh M-CSFR/CSF1R; Catalog#10161-H08H; NP_005202.2; Met1-Glu512). M-CSFR/CSF1R specific IgG was purified by Human M-CSFR/CSF1R affinity chromatography.
Catalog No. 50-255-5451