Learn More
Sino Biological Human CSF1R PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10161T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CSF1R (rh CSF1R; Catalog#10161-H08H; NP_005202.2; Met1-Glu512). CSF1R specific IgG was purified by Human CSF1R affinity chromatography.
Catalog No. 50-237-2844