Learn More
Sino Biological Human CSF1R Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10161H49HB20
Encompass_Preferred
A DNA sequence encoding the human CSF1R (NP_005202.2)(Met1-Glu512) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2153