Learn More
Sino Biological Anti-GM-CSF/CSF2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10015MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GM-CSF / CSF2 (rh GM-CSF / CSF2; Catalog#10015-H07H; NP_000749.2; Ala18-Glu144). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8509