Learn More
Sino Biological Human CSF2 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10015R02720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human GM-CSF / CSF2 (rh GM-CSF / CSF2; Catalog#10015-H07H; NP_000749.2; Ala18-Glu144).
Catalog No. 50-237-3746