Learn More
Sino Biological Recombinant Anti-GM-CSF Receptor alpha Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10701R009F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD116 / GM-CSFR (rh CD116 / GM-CSFR; Catalog#10701-H08H; P15509-1; Met1-Gly320) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1604