Learn More
Sino Biological Human CSF2RA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10701T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GM-CSF Receptor alpha (rh GM-CSF Receptor alpha; Catalog#10701-H08H; P15509-1; Met1-Gly320). GM-CSF Receptor alpha specific IgG was purified by Human GM-CSF Receptor alpha affinity chromatography.
Catalog No. 50-237-2432