Learn More
Sino Biological Anti-IL-3R Beta/IL3RB Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10516MM01A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD131 / CSF2RB / IL3RB / IL5RB (rh CD131 / CSF2RB / IL3RB / IL5RB; Catalog#10516-H08H; NP_000386.1; Met1-Trp443) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2133