Learn More
Sino Biological Anti-IL-3R Beta/IL3RB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10516RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CSF2RB (rh CSF2RB; Catalog#10516-H08H; NP_000386.1; Met 1-Trp 443). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1508