Learn More
Sino Biological Anti-G-CSF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10007RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GCSF (rh G-CSF; Catalog#10007-HNCE; NP_757373.1; Ala 30-Pro 204). G-CSF specific IgG was purified by human G-CSF affinity chromatography.
Catalog No. 50-240-8751