Learn More
Sino Biological Anti-CSF3R/G-CSFR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10218MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human G-CSFR extracellular domain (rhG-CSFR; Catalog #10218-HCCH; Met1-Pro621; NP_000751.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1019