Learn More
Sino Biological Anti-Placental Lactogen/CSH1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11596RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CSH1 (rh CSH1; Catalog#11596-H08H; NP_001308.1; Met 1-Phe 217). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4685