Learn More
Sino Biological Recombinant Human Placental Lactogen/CSH1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11596H08HB20
Encompass_Preferred
A DNA sequence encoding the human CSH1 (NP_001308.1) (Met1-Phe217) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4686