Learn More
BIOHIPPO Human CSN3 Microsample
SDP
Catalog No. 5000314900
Encompass
Human CSN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CSN3 in Breast milk and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 34 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CSN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CSN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CSN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14900 Supplier BIOHIPPO Supplier No. BHE15211440
Human CSN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CSN3 in Breast milk and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 34 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CSN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CSN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CSN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color