Learn More
Abcam Human CSPG4 ELISA Kit (NG2/MCSP), 1 x 96 Tests
Catalog No. 502656460
Encompass_Preferred
Human CSPG4 ELISA Kit (NG2/MCSP) is a single-wash 90-min Simplestep used to quantify Human CSPG4 (NG2/MCSP) with a sensitivity of 25.5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6460 Supplier Abcam Supplier No. AB2678071X96TESTS
Human CSPG4 ELISA Kit (NG2/MCSP) is a single-wash 90-min Simplestep used to quantify Human CSPG4 (NG2/MCSP) with a sensitivity of 25.5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement