Learn More
Sino Biological Anti-Cystatin C Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10439MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cystatin C / CST3 (rh Cystatin C / CST3; Catalog#10439-H08H; NP_000090.1; Ser27-Ala146). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8355