Learn More
Sino Biological Human CST4 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11542MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cystatin S / CST4 (rh Cystatin S / CST4; Catalog#11542-H08H; NP_001890.1; Met 1-Ala 141). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1558