Learn More
Sino Biological Human CST5 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10506RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cystatin D / CST5 (rh CST5; Catalog#10506-H08H; NP_001891.2; Met1-Val142). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1849