Learn More
Sino Biological Anti-CST6 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10438MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cystatin-M / CST6 (rh Cystatin-M; Catalog#10438-H08H; NP_001314.1; Met 1-Met 149). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0583