Learn More
BIOHIPPO Human CST7 Microsample
SDP
Catalog No. 5000313782
Encompass
Human CST7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CST7 in Serum plasma cell culture supernates It provides a detection range of 0 625-40 ng/mL with a sensitivity of 0 152 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CST7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CST7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CST7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13782 Supplier BIOHIPPO Supplier No. BHE15212452
Human CST7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CST7 in Serum plasma cell culture supernates It provides a detection range of 0 625-40 ng/mL with a sensitivity of 0 152 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CST7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CST7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CST7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color