Learn More
Sino Biological Anti-Cystatin F/CST7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10167T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cystatin F/CST7 (rh Cystatin F/CST7; Catalog#10167-H08H; NP_003641.3; Met1-His145). Cystatin F/CST7 specific IgG was purified by Human Cystatin F/CST7 affinity chromatography.
Catalog No. 50-255-5446