Learn More
Sino Biological Human CSTA MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10479R236100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin A / CSTA (rh Cystatin A / CSTA; Catalog#10479-H07E; P01040; Ile2-Phe98).
Catalog No. 50-237-2667