Learn More
Sino Biological Anti-Cystatin A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10479T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cystatin A / CSTA (rh Cystatin A / CSTA; Catalog#10479-H07E; P01040; Ile2-Phe98). Cystatin A / CSTA specific IgG was purified by Human Cystatin A / CSTA affinity chromatography.
Catalog No. 50-240-8936