Learn More
Sino Biological Anti-Cystatin B Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10435MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cystatin B / CSTB (rh Cystatin B / CSTB; Catalog#10435-H07E; P04080; Met 2-Phe 98). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0779