Learn More
Biomatik Corporation Human C-Terminal Binding Protein 2 (CTBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU03515
Encompass_Preferred
Human C-Terminal Binding Protein 2 (CTBP2) ELISA Kit for detecting CTBP2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56545. Gene ID: 1488. Target Synonyms: Ribeye. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9028