Learn More
Sino Biological Human CTF1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 16013H01H1
Encompass_Preferred
A DNA sequence encoding the human CTF1(NP_001321.1)(Ser2-Ala201) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-0602