Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CTHRC1 ELISA Kit

Catalog No. EH135RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH135RBX5 5 x 96 Tests
EH135RBX10 10 x 96 Tests
EH135RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH135RBX5 Supplier Invitrogen™ Supplier No. EH135RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human CTHRC1 quantitates human CTHRC1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CTHRC1.

The Collagen triple helix repeat-containing protein (CTHRC1) is a glycosylated, secreted protein that is transiently expressed in the arterial wall in response to injury where it is thought to contribute to vascular remodeling by inhibiting collagen expression and deposition and promoting cell migration. CTHRC1 is highly expressed multiple human cancers, including pancreatic cancer where it plays a significant role in the progression and metastasis of the disease by regulating migration and adhesion of tumors cells. CTHRC1 overexpression can also be used as an independent prognostic marker in gastric cancer. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96CG8
Assay Range 8-5000 pg/mL
Assay Sensitivity 8 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110014B07Rik; collagen triple helix repeat containing 1; collagen triple helix repeat-containing protein 1; CTHRC1; Protein NMTC1; UNQ762/PRO15501110014B07Rik; collagen triple helix repeat containing 1; collagen triple helix repeat-containing protein 1; CTHRC1; Protein NMTC1; UNQ762/PRO1550;
Gene ID (Entrez) 115908
Gene Symbol Cthrc1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CTHRC1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.